Learn More
Meso Scale Discovery UPLEX NHP IL2 ASY384well
SDP
Supplier:  Meso Scale Discovery K256TVK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-2 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions It allows for the accurate quantification of natural human IL-2 levels in just 3 5 hours using as little as 25 L of sample
Catalog No. 50-000-81230
Add to Cart
Request a Quote