Learn More
Meso Scale Discovery UPLEX NHP IL23 ASY384well
SDP
Supplier:  Meso Scale Discovery K256WGK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-23 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human IL-23 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve It allows efficient and accurate quantification of IL-23 levels in samples The kit has a sensitivity of 0 04 pg/mL and requires only 12 5 L of sample per well
Catalog No. 50-000-78425
Only null left
Add to Cart
Request a Quote