Learn More
Meso Scale Discovery UPLEX NHP IL5 ASY384well
SDP
Supplier:  Meso Scale Discovery K256UOK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-5 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring IL-5 in human non-human primate rat and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls Requiring only 25 L of sample it quantifies IL-5 from 0 05-1000 pg/mL with a dynamic range of up to 4 logs
Catalog No. 50-000-81186
Only null left
Add to Cart
Request a Quote