Learn More
Meso Scale Discovery UPLEX NHP MCP1 ASY384well
SDP
Supplier:  Meso Scale Discovery K256UGK4
Encompass
The Meso Scale Discovery U-PLEX NHP MCP-1 384-well Assay Kit is a multiplex immunoassay that measures human monocyte chemoattractant protein 1 (MCP-1) in serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has high sensitivity wide dynamic range and requires low sample volume This product can aid research on inflammatory biomarkers like MCP-1
Catalog No. 50-000-81372
Add to Cart
Request a Quote