Learn More
Meso Scale Discovery UPLEX NHP MCP2 ASY384well
SDP
Supplier:  Meso Scale Discovery K256XHK4
Encompass
The U-PLEX NHP MCP-2 (384-well assay) K256XHK from Meso Scale Diagnostics is a 384-well multiplex assay kit that enables simultaneous measurement of up to 10 biomarkers per well It uses electrochemiluminescence (ECL) detection technology and is designed for use on Meso Scale Diagnostics imaging systems This particular kit includes reagents for measuring human MCP-2 It requires just 25 L of sample per well and the small assay volume is enabled by the combination of ECL technology and plate design The kit includes a 384-well high bind ECL plate detection antibodies read buffer sample diluent calibrators and controls
Catalog No. 50-000-81770
Add to Cart
Request a Quote