Learn More
Meso Scale Discovery UPLEX NHP MIP3 ASY384well
SDP
Supplier:  Meso Scale Discovery K256UZK4
Encompass
The MIP-3alpha Human ELISA Kit from Meso Scale Discovery is an enzyme-linked immunosorbent assay for the quantitative measurement of MIP-3alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody standards diluent read buffer wash buffer substrate and stop solution The assay has a sensitivity of 0 3 pg/mL and a detection range of 0 49-1500 pg/mL It uses electrochemiluminescence for high sensitivity and wide dynamic range The total assay time is 3 5 hours This product can be used to measure MIP-3alpha levels in immune response studies
Catalog No. 50-000-80218
Add to Cart
Request a Quote