Learn More
Meso Scale Discovery UPLEX NHP TGFB3 ASY384well
SDP
Supplier:  Meso Scale Discovery K256XVK4
Encompass
The Meso Scale Discovery U-PLEX NHP TGF beta 3 384-well assay is a multiplex immunoassay that measures active and total TGF beta 3 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This 384-well plate assay allows high throughput screening and includes all necessary reagents for performing the immunoassay
Catalog No. 50-000-78429
Add to Cart
Request a Quote