Learn More
Meso Scale Discovery UPLEX NHP YKL40 ASY384well
SDP
Supplier:  Meso Scale Discovery K256VLK4
Encompass
The Meso Scale Discovery U-PLEX NHP YKL-40 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring YKL-40 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective assay The detection range is 1 15 - 10 000 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators controls 10X read and wash buffers and plate seals enabling high-throughput quantification with minimal sample volume
Catalog No. 50-000-80264
Only null left
Add to Cart
Request a Quote