Learn More
Meso Scale Discovery UPLEX Oncolysis Combo 1HMN
SDP
Supplier:  Meso Scale Discovery K15603K4
Encompass
The U-PLEX Oncolysis Combo 1 Human Sector K15603K by Meso Scale Diagnostics is a multiplex immunoassay kit for measuring 10 oncolysis-related biomarkers It includes a 96-well plate with capture and detection antibodies Read Buffer Diluent 2 Calibrator Blend and Controls It detects granzyme B HMGB1 sFasL TNF-alpha TRAIL IL-6 IL-8 MCP-1 MIF and PAI-1 from 50 L samples using electrochemiluminescent detection The kit tests 40 samples in duplicate with standard curves and controls intended for research use only
Catalog No. 50-000-80309
Add to Cart
Request a Quote