Learn More
Meso Scale Discovery UPLEX Proinflam Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15049K4
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures 10 human inflammatory biomarkers (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay allows efficient quantification of multiple inflammatory cytokines from a single small-volume sample
Catalog No. 50-000-79186
Add to Cart
Request a Quote