Learn More
Meso Scale Discovery UPLEX Rat CPeptide ASY384well
SDP
Supplier:  Meso Scale Discovery K2536JK2
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide 384-Well Assay is a multiplex immunoassay kit for the quantitative measurement of C-Peptide in rat serum plasma and cell culture supernatants The assay utilizes electrochemiluminescent detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The lower limit of detection is 15 6 pg/mL This assay allows for the accurate quantification of C-Peptide levels in rat biological samples
Catalog No. 50-000-81159
Add to Cart
Request a Quote