Learn More
Meso Scale Discovery UPLEX Rat FGF21 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K1535WK24
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay that measures FGF-21 levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires 50 L of sample per well and can assay 46 samples in duplicate The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78269
Only null left
Add to Cart
Request a Quote