Learn More
Meso Scale Discovery UPLEX Rat Ghrelin act ASYQP
SDP
Supplier:  Meso Scale Discovery K1536KK24
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit quantifies active ghrelin in rat serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls diluent read buffer wash buffer and seals Sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL Using the QuickPlex SQ 120 results are available in under 3 hours enabling rapid and accurate quantification of active ghrelin
Catalog No. 50-000-81364
Only null left
Add to Cart
Request a Quote