Learn More
Meso Scale Discovery UPLEX Rat Ghrelin active ASY
SDP
Supplier:  Meso Scale Discovery K1536KK4
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin levels in rat serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of active ghrelin and total ghrelin from a single sample The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators controls 10X Read Buffer 10X Wash Buffer Blocker A and Blocker D The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-1000 pg/mL
Catalog No. 50-000-79757
Add to Cart
Request a Quote