Learn More
Meso Scale Discovery UPLEX Rat GLP1 inactive ASY
SDP
Supplier:  Meso Scale Discovery K1535VK1
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) Assay Kit measures GLP-1 (7-36) amide and GLP-1 (9-36) amide in rat serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager instruments with an assay time of 3 5 hours for 96 samples offering a high-throughput method for measuring inactive GLP-1
Catalog No. 50-000-81516