Learn More
Meso Scale Discovery UPLEX Rat GLPactive ASY384well
SDP
Supplier:  Meso Scale Discovery K2536LK2
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and has a detection range of 0 61-2500 pg/mL with an LLOQ of 0 61 pg/mL The kit includes a 384-well carbon electrode plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The total assay time is 3 5 hours This assay allows high-throughput quantification of active GLP-1
Catalog No. 50-000-80198
Only null left
Add to Cart
Request a Quote