Learn More
Meso Scale Discovery UPLEX Rat GLPinactive ASY384
SDP
Supplier:  Meso Scale Discovery K2535VK2
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) 384-Well Assay Kit is a multiplex immunoassay that measures inactive GLP-1 in rat serum plasma and cell culture supernatants This 384-well plate assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies and calibrators and controls to generate a standard curve The assay has a typical lower limit of detection of 1 23 pg/mL and an upper limit of quantitation of 10 000 pg/mL for rat GLP-1 (inactive)
Catalog No. 50-000-78974
Only null left
Add to Cart
Request a Quote