Learn More
Meso Scale Discovery UPLEX Rat GLPinactive ASY384
SDP
Supplier:  Meso Scale Discovery K2535VK4
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Inactive) 384-Well Assay Kit measures inactive GLP-1 in rat serum plasma and cell culture supernatants This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the Meso QuickPlex SQ 120 The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours offering high sensitivity and a broad dynamic range for robust measurement of inactive GLP-1
Catalog No. 50-000-81219
Add to Cart
Request a Quote