Learn More
Meso Scale Discovery UPLEX Rat Insulin ASYQP
SDP
Supplier:  Meso Scale Discovery K1536HK24
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin Assay Kit is an electrochemiluminescent immunoassay for measuring rat insulin in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to 0 05 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and sample diluent for serum EDTA plasma heparin plasma and cell culture supernatants It takes 3 5 hours and is for use on MSD platforms
Catalog No. 50-000-78584
Only null left
Add to Cart
Request a Quote