Learn More
Meso Scale Discovery UPLEX TBI Combo 1 human
SDP
Supplier:  Meso Scale Discovery K15685K4
Encompass
The Meso Scale Discovery U-PLEX TBI Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures biomarkers related to traumatic brain injury (TBI) in human serum plasma and CSF samples The kit includes reagents to measure GFAP UCH-L1 S100B and Tau in a single well of a 96-well plate using electrochemiluminescence detection This allows efficient and accurate quantification of these important TBI biomarkers The kit has high sensitivity a broad dynamic range and requires a low sample volume It provides a streamlined workflow to profile TBI biomarkers from human biofluids
Catalog No. 50-000-81035
Only null left
Add to Cart
Request a Quote