Learn More
Meso Scale Discovery UPLEX TH1/TH2 Combo NHP
SDP
Supplier:  Meso Scale Discovery K15080K4
Encompass
The U-PLEX TH1/TH2 Combo NHP Assay is a multiplex immunoassay panel that simultaneously measures 10 biomarkers related to T helper cell activation in non-human primate samples It uses electrochemiluminescence detection technology on the Meso Scale Discovery platform to quantitatively measure IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A TNF- and GM-CSF from serum plasma and culture supernatants This assay allows researchers to profile non-human primate samples for TH1/TH2 immune responses and inflammation It requires 50 L of sample per well and the total assay time is approximately 5 hours
Catalog No. 50-000-81598
Add to Cart
Request a Quote