Learn More
Electron Microscopy Sciences Upright Compound Microscope Dual View, Achromatic
SDP
Supplier:  Electron Microscopy Sciences CP300DH
Encompass_Preferred
Bright field microscope with dual optical body allows two people to observe simultaneously, inclined 45 degree, and able to rotate a full 360 degree with 55-75 mm interpupillary adjustment. Left eye diopter adjustment of -5 to +5. Quadruple ball-bearing nosepiece with positive click stops and smooth operation. Eyepieces are 10X wide field with a focal length of 25 mm, Field F 18 mm.
Stage 140 mm X 135 mm withrange of motion of 75 mm X 50, integrated mechanical stage with vernier, low position coaxial control knobs, and removable spring clip slide holder.
Focus mechanism with all stainless steel and brass gear train system. Adjustable tension control to prohibit drift. Adjustable up-stop to protect slides, specimen, and lenses. Coaxial coarse and fine adjustments with dial markings at 0.002 mm increments.
Includes mold protection throughout the optical body, mirror assembly, dust cover, immersion oil, two color filters, spare halogen lamp, and spare fuse.
Catalog No. 50-343-61