Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Stainless Stainless Freezer Rack, (2) Drawers, Holds (60) 15ml + (30) 50ml Tubes Rack Dimensions 16 11/16L x 9 15/16H x 5 1/2W" (424 x 252 x 140mm)
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFDT15/50
Encompass_Preferred
Rack Designed to hold (60) 15ml Tubes in 1 Drawer + (30) 50ml Centrifuge Tubes in 1 Drawer, made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
Catalog No. NC2208067