Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Style Freezer Rack, Holds (12) Large 3.75" Boxes, Rack Dims 18 7/16L x 16 11/16H x 6 1/8W" (459 x 424 x 155mm) Compartment Dims 5 7/8L x 3 15/16H x 5 13/16W" (149 x 100 x 148mm) 3x4
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFD34L375
Encompass_Preferred
Rack Designed to hold (12) Large 3.75H"Cardboard Boxes ((3 7/8H x 5 13/16L x 5 13/16W", (98 x 146 x 146mm)) made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
Catalog No. NC2208083