Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Drawer Freezer Rack, Holds (21) 100-Place Slide Boxes, Rack Dims 22 3/16L x 12 1/16 x 8 5/8W" (564 x 307 x 219mm), with Compartment Dims 7 1/8L x 1 1/2H x 8 1/4W" (181 x 38 x 210mm) 3x7
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFDS10037
Encompass_Preferred
Rack Designed to hold (21) 100 Place Slide Boxes (8.25H x7W x1.34L" (210 x 178 x 34mm), made of corrosion resistant stainless steel, providing access to samples without having to remove the whole rack, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
Catalog No. NC2208113