Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Freezer Rack, Mixed Use Storage, Various Tubes, Bottles, Boxes, Rack Dims 22 1/16L x 11H x 5 7/16W" (560 x 280 x 138mm) Drawer Area 21 9/16 x 10 15/16 x 5 3/16" (548 x 278 x 132mm)
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFTM115
Encompass_Preferred
Rack Designed to store a variety of tubes,bottles, Boxes etc. in 1 Bin, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack. Great for organizing -20⁰, -30⁰ , -86⁰ Freezers and Cold Rooms
Catalog No. NC2208091