Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Bin Rack with Lid, Large, 26.54" x 6.69" x 5.43" (L x H x W)
Supplier:  Cole-Parmer 0439783
Encompass_Preferred
Available in a wide variety of sizes, the PolarSafe stainless steel bin storage racks are designed to store miscellaneous items such as bottles, tubes, vials, bags or boxes that will not fit in a standard storage rack. The racks feature wire mesh sides for easy viewing of contents and dual handles for carrying. They are available in fixed or drawer rack styles which are fully framed.
Catalog No. NC1686853