Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Combination Drawer Racks for15 & 50ml Centrifuge Tubes (60 )15ml & (30) 50ml Tubes
SDP
Supplier:  Alkali Scientific UFDRT15/50
Encompass
For 15/50 mL Centrifuge Tubes Upright Combination Drawer Freezer Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks hold 60 x 15ml and 30 x 50ml centrifuge tubes Security lock devices available for each rack Dimensions L x H x W: 16 11/16 x 9 15/16 x 5 1/2 inches (424 x 252 x 140mm)Capacity: 15 mL: 60 Tubes/ 50mL: 30 Tubes*Lock device is ordered separately.
Catalog No. 50-302-6707