Learn More
Labrepco Inc UPRIGHT FREEZER COMBINATION DR
SDP
Supplier:  Labrepco Inc UFDT15322
Encompass_Preferred
Combination drawer rack, Upright freezer; Labrepco; Crystal; For 2 in. boxes, 15mL and 50mL Centrifuge tubes; Capacity: 6 Boxes plus 60 Tubes (15mL); L x H x D: 282 x 246 x 140mm
Catalog No. 50-902-0117