Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 15ml Centrifuge Tubes – Rack Only (2 Drawers; 120 Tubes)
SDP
Supplier:  Alkali Scientific UFDRT152
Encompass_Preferred
For 15ml Centrifuge Tubes Upright Freezer Drawer Rack stores 15 ml Centrifuge Tubes and features 2 drawers for storage up to 120 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer.   Dimensions (DxWxH): 16 11/16 x 9 15/16 x 5 1/2  inches (424 x 252 x 140mm)Capacity: 120 Tubes *Lock device is ordered separately.
Catalog No. 50-302-6708