Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 50mL Centrifuge Tubes – Rack Only (1 Drawers, 39 Tubes)
SDP
Supplier:  Alkali Scientific UFDRLT501
Encompass_Preferred
For 50ml Centrifuge Tubes Upright Freezer Drawer Rack stores 50 ml Centrifuge Tubes and features 1 drawer for storage up to 39 tubes. These Upright Freezer Drawer Racks are specifically designed to provide an ideal alternative for storing large quantities of samples in centrifuge tubes. Constructed of corrosion resistant stainless steel, these freezer racks include handles and identification card holders. Drawer racks facilitate easy access of samples without having to remove the whole rack from the freezer. Dimensions (DxWxH): 22 3/16 x  4 15/16 x 5 1/2  inches (564 x 126 x 140mm)Capacity: 39 Tubes *Lock device is ordered separately.
Catalog No. 50-302-7820