Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for 15ml and 50ml Centrifuge Tubes, 80 Tubes 15mL and 39 Tubes 50mL
SDP
Supplier:  Alkali Scientific UFDLT15/50
Encompass
Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Constructed of corrosion resistant stainless steel Racks hold 80 x 15ml and 39 x 50ml centrifuge tubes Security lock devices available for each rack Dimensions L x H x W: 22 3/16 x 9 15/16 x 5 1/2 inches (564 x 252 x 140mm) Capacity: 15 mL: 80 Tubes/ 50mL: 39 Tubes *Lock device is ordered separately.  
Catalog No. 50-302-8013