Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Rack for Blood Sample Tubes – Fits 3mL Tubes, 270 Capacity, 3 Drawers
SDP
Supplier:  Alkali Scientific UFDRT033
Encompass
Upright Freezer Drawer Rack for storage of Blood Sample Tubes allows for storage of up to 270 tubes for 3ml sizes. This freezer rack features three drawers and is made with corrosion resistant stainless steel.  Freezer drawer racks include handles, identification card holders and conveniently allow for storage & tube retrieval without removing the whole rack from the freezer.  Optional security lock devices are also available for each rack, sold separately. Dimensions (DxWxH): 16 11/16 x 5 1/2 x 9 1/8 inches (424 x 140 x 231mm)Bottom of Form
Catalog No. 50-302-7841