Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Drawer Racks for Standard 3.75″ Boxes- Rack Only- 5 Boxes Deep x 2 Boxes High
SDP
Supplier:  Alkali Scientific UFR52375
Encompass_Preferred
For 3.75″ High Standard Boxes These Upright Freezer Drawer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Dimensions L x H x W: 27 3/16 x 8 1/16 x 5 1/2 inch (691 x 205 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm
Catalog No. 50-302-8004