Learn More
Thomas Scientific Rack with 4 drawers (6 compartments per drawer) for Microtiter Plates
SDP
Supplier:  Thomas Scientific 1148W99
Encompass_Preferred
Racks are for 96-well & 384-well Microtiter Plates. Drawer Racks allow an easy access to fewer Microtiter plates at a time thus protecting other samples in the rack from warming up. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-045