Learn More
Chemglass Life Sciences Freezer Rack Only, Upright Drawer Style, for Standard 2" Boxes, Box Capacity: 3L x 4H, 16-1/2"L x 5-1/2"W x 9-7/16"H
SDP
Supplier:  Chemglass Life Sciences CLS3795003
Encompass
Freezer Rack Only, Upright Drawer Style, for Standard 2" Boxes, Box Capacity: 3L x 4H, 16-1/2"L x 5-1/2"W x 9-7/16"H. High quality racks are constructed of corrosion-resistant stainless steel and include handles and identification card holders.
Catalog No. NC1703138