Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Freezer Rack, Holds (6) Centrifuge Boxes, Rack Dimensions 18 13/16L x 10 3/16H x 5 7/8W" (478 x 259 x 149mm) with Compartment Dimensions 6L x 5 1/16H x 5 13/16W" (152 x 128 x 147.5mm) 3 x 2
Supplier:  CRYSTAL TECHNOLOGY AND INDUSTRIES UFLB32
Encompass_Preferred
Rack Designed to hold (6) 5"H 15ml and 50ml Centrifuge Tube Boxes, (4 15/16H x 5 13/16L x 5 13/16W" (152 x 148 x 148mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
Catalog No. NC2208040