Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 2″ Box Storage in PHCbi & NuAire Lab Freezers; Rack Only; 4x5 Configuration, 20 Box Capacity
SDP
Supplier:  Alkali Scientific UFDR452S
Encompass_Preferred
Panasonic Specific Model: Upright freezer racks for storage of 2-inch boxes, features a 4 boxes deep by 5 boxes high configuration for a total storage of 20 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. This drawer style freezer rack does not come standard with storage boxes. This is a custom-built rack for certain 25 Cu. Ft. freezers from PHCbi (formerly Panasonic/Sanyo) & NuAire. Dimensions (D x H x W): 22 7/16 x 11 9/16 x 5 1/2 inches (566 x 293 x 140mm)Compartment Dimensions: 5 3/8 x 2 1/16 x 5 1/4 inches (137.4 x 52 x 133mm)
Catalog No. 50-302-8023