Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 32/8 Flipper JR Reversible Multi-Boxes, 6 x 4 Configuration, 24 Box Capacity
SDP
Supplier:  Alkali Scientific UFF64
Encompass_Preferred
Easily access boxes from either side of rack with right or left handed doors. Our freezer racks have been configured to fit all major ultra low (-80 deg) freezers on the market; racks are designed for ultra low upright freezers. Made of corrosion resistant stainless steel Dimensions (DxWxH): 21 1/2 x 6 15/16 x 9 3/16 inches (546 x 177 x 233mm)Compartment Dimensions: 3 7/16 x 6 7/8 x 2 3/16 inches (87 x 174 x 55mm)
Catalog No. 50-302-7869