Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 50-Cell 1.5ml Microtube Storage Boxes, 1x4x2 Configuration, 8 Box Capacity
SDP
Supplier:  Alkali Scientific UFM1415
Encompass_Preferred
For 50-cell 1.5mL Storage Boxes The UFM-1415 Upright Freezer Racks are designed for storage of 50-Cell 1.5ml Microtube Boxes and feature a 4 Boxes High x 2 Boxes Deep configuration. These upright freezer racks include handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format.  Each rack compartment holds two 50 Cell 1.5ml Microtube Storage Boxes. Microtube boxes are not included with purchase. Security devices are available for each rack though ordered separately. Dimensions (DxWxH): 6 3/4 x 5 7/8 x 9 1/4 inches (172 x 149 x 235mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147 x 57.6mm
Catalog No. 50-302-7990