Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 3.75″ Boxes- Rack Only- 3 Boxes Deep x 2 Boxes High
SDP
Supplier:  Alkali Scientific UFR32375
Encompass_Preferred
For 3.75″ High Standard Boxes Standard upright freezer racks for storage of 3.75 inch boxes, features a 3 boxes deep by 2 boxes high configuration for a total storage of 6 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 16 13/16 x 8 1/16 x 5 1/2 inch (427 x 205 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm
Catalog No. 50-302-8101