Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 3.75″ Boxes- Rack Only- 2 Boxes Deep x 3 Boxes High
SDP
Supplier:  Alkali Scientific UFR23375
Encompass_Preferred
For 3.75″ High Standard BoxesCapacity Box – 2 x 3 =6 Standard upright freezer racks for storage of 3.75-inch boxes, features 2 boxes deep by 3 boxes high configuration for a total storage of 6 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 11 3/8 x 12 1/16 x 5 1/2 inch (289 x 307 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)
Catalog No. 50-302-8516