Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 96-Well and 384-Well Plates with Locking Rod – Rack Only (168-204 Plates)
SDP
Supplier:  Alkali Scientific UFRMP619L
Encompass_Preferred
For 96-well & 384- Well Microtiter Plates Upright Freezer Racks are designed for storage of 96 & 384-Well Microtiter Plates and can store up to 80 plates. These freezer racks are constructed of corrosion resistant stainless steel, include handles, and offer convenient storage of microtiter plates in your laboratory freezer.  Locking rods are also included for added security. Microtiter plates are not included with purchase. Plate Capacity: 168 with lids 204 without lids Dimensions (DxWxH): 22 1/2 x 5 1/2 x 18 15/16 inches (571 x 140 x 481mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 9 7/16 inches (91 x 130.5 x 239mm)
Catalog No. 50-302-7852