Learn More
Labrepco Inc UPRIGHT FREEZER RACKS FOR STD
SDP
Supplier:  Labrepco Inc UF362
Encompass_Preferred
Rack, Upright freezer; Labrepco; Crystal; For standard 2 in. boxes; Capacity (Box): 18; L x H x D: 415 x 321 x 140mm
Catalog No. 50-902-0096