Learn More
Alkali Scientific Alkali Scientific™ Upright Sliding Tray Freezer Rack for 2" Boxes, 5 x 5 Configuration, 25 Box Capacity
SDP
Supplier:  Alkali Scientific UFS552
Encompass_Preferred
For Standard 2″ Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions (LxHxW): 27 1/16 x 11 x 5 1/2 (688 x 280 x 140mm)Compartment Dimensions: 5 5/16 x 2 1/16 x 5 1/4 (134.1 x 53 x 134.1mm)
Catalog No. 50-302-8127