Learn More
Alkali Scientific Upright Stainless-Steel Freezer Rack for 2" Boxes, 3 x 5 Configuration, 15 Box Capacity
SDP
Supplier:  Alkali Scientific UF352
Encompass
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks fit standard 2" cardboard, plastic, or metal boxes. Dimensions (DxHxW): 16 7/8 x 11 x 5 1/2 inches (429 x 280 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm).
Catalog No. 50-238-9041