Learn More
Enterprise Technology Solutions CyberPower PR1500RT2UCN Smart App Sinewave Rack/Tower Convertible UPS
SDP
Supplier:  Enterprise Technology Solutions PR1500RT2UCN
Encompass_Preferred
CyberPower brings high-tech power protection and battery backup together with the PR1500RT2UCN Smart App Sinewave Rack/Tower Convertible UPS. This UPS system is versatile as it can be installed vertically or horizontally, which allows for easier accommodation of devices connected to the eight NEMA 5-15R outlets. If power loss occurs, you can properly shut down your network devices as all outlets can receive up to 6.5 minutes of power.
Catalog No. 50-001-72695