Learn More
Medchemexpress LLC Uracil-13C4 | 66-22-8 | 98.0% | 116.04 g/mol | 13C4H4N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYI0960S110MG
Encompass_Preferred
Uracil-13C4 is the 13C-labeled form of uracil, a pyrimidine nucleobase found in RNA. The isotopically labeled compound is supplied for use as a reference standard or tracer in analytical and metabolic studies that require isotope-labeled material.
  • 13C-labeled at four carbon positions
  • High purity (98.0%)
  • Formula: 13C4H4N2O2; molecular weight: 116.04 g/mol
  • Supplied as a 10 mg vial suitable for analytical workflows
Catalog No. 50-002-97607