Learn More
Medchemexpress LLC Uradil impurity 1 | 31737-09-4 | 99.8% | 160.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY20821100MG
Encompass_Preferred
Uradil impurity 1 is an impurity of Uradil, intended for research use only. MedChemExpress does not sell to patients.
  • Soluble in DMSO at 100 mg/mL
  • Stock solutions stored at -80°C should be used within 6 months
  • Stock solutions stored at -20°C should be used within 1 month
Catalog No. 50-004-01049