Learn More
Electron Microscopy Sciences Uranyl acetate, 6159-44-0, MFCD00150376, 25g
SDP
Supplier:  Electron Microscopy Sciences 22400
Encompass_Preferred
Molecular Formula C4H6O6U.2H2O, Molecular Weight 424.145; Uranyl Acetate Dihydrate: A universal EM stain for thin sections, en-block staining, and negative staining.
Catalog No. NC0788109
This product is regulated by the federal government, and may not be ordered online.